Trifluoroacetic acid-[D₁] ≥99%

Supplier: Matrix Scientific

086141-1G 086141-10G
102847-078EA 56.92 USD
102847-078 102847-080
Trifluoroacetic acid-[D₁] ≥99%
Trifluoroacetic acid-[D₁]

Hazard: Irritant

Formula: F₃CCOOD
MDL Number: MFCD00044563
CAS Number: 599-00-8
UN: 2699
ADR: 8,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR