N-[2-(Dimethylamino)ethyl]-N-methylguanidinehydroiodide ≥97%

Supplier: Matrix Scientific

086125-1G 086125-10G
102847-026EA 156.92 USD
102847-026 102847-028
N-[2-(Dimethylamino)ethyl]-N-methylguanidinehydroiodide ≥97%
N-[2-(Dimethylamino)ethyl]-N-methylguanidinehydroiodide

Hazard: Irritant

Formula: C₆H₁₇IN₄
MW: 272.13 g/mol
Melting Pt: 107…113 °C
MDL Number: MFCD06797493
CAS Number: 849776-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR