2,6-Dimethyl-3,5-heptanedione ≥97%
Supplier: Matrix Scientific
086123-1G
086123-10G
102847-018EA
101.54
USD
102847-018
102847-020
2,6-Dimethyl-3,5-heptanedione ≥97%
2,6-Dimethyl-3,5-heptanedione
Hazard: Irritant
Formula:
C₉H₁₆O₂ MW: 156.22 g/mol |
MDL Number:
MFCD00015040 CAS Number: 18362-64-6 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...