Trimethyl orthovalerate ≥97%
Supplier: Matrix Scientific
086119-1G
086119-10G
086119-50G
102847-004EA
44.62
USD
102847-004
102847-006
102847-008
Trimethyl orthovalerate ≥97%
Trimethyl orthovalerate
Hazard: Irritant
Formula:
C₈H₁₈O₃ |
MDL Number:
MFCD00008481 CAS Number: 13820-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...