2-Amino-2-thioxoethyl pivalate ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Amino-2-thioxoethyl 2,2-dimethylpropanoate
086117-1G
086117-10G
102846-994EA
184.62
USD
102846-994
102846-996
2-Amino-2-thioxoethyl pivalate ≥97%
2-Amino-2-thioxoethyl pivalate
Hazard: Irritant
Formula:
C₇H₁₃NO₂S MW: 175.25 g/mol |
MDL Number:
MFCD00204238 CAS Number: 175204-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...