Octadecyltrimethylammonium bromide ≥98%
Supplier: Matrix Scientific
Synonyms:
OTAB, STAB, Trimethylstearylammonium bromide, OctMAB, Trimethyloctadecylammonium bromide, N,N,N-Trimethyloctadecan-1-aminium bromide, Octadecyltrimethylammonium bromide, Stearyltrimethylammonium bromide
086115-10G
086115-50G
102846-986EA
129.23
USD
102846-986
102846-988
Octadecyltrimethylammonium bromide ≥98%
Octadecyltrimethylammonium bromide
Hazard: Irritant
Formula:
C₂₁H₄₆BrN MW: 392.5007 g/mol |
MDL Number:
MFCD00043171 CAS Number: 1120-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...