Methylsulfonylacetonitrile ≥97%

Supplier: Matrix Scientific

086104-1G 086104-10G 086104-50G
102846-946EA 38.46 USD
102846-946 102846-948 102846-950
Methylsulfonylacetonitrile ≥97%
Methylsulfonylacetonitrile

Hazard: Toxic

Formula: C₃H₅NO₂S
MW: 119.14 g/mol
Melting Pt: 81…86 °C
MDL Number: MFCD00014743
CAS Number: 2274-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR