Sodium diethyldithiocarbamate 25% in aqueous solution , 97%
Supplier: Matrix Scientific
086103-10G
086103-50G
102846-942EA
92.31
USD
102846-942
102846-944
Sodium diethyldithiocarbamate 25% in aqueous solution , 97%
Sodium diethyldithiocarbamate
Hazard: Irritant
Formula:
C₅H₁₀NNaS₂ MW: 171.26 g/mol |
CAS Number:
148-18-5 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...