3,3-Dimethyl-1-(trimethylsilyl)-1-butyne ≥97%
Supplier: Matrix Scientific
086100-1G
086100-10G
102846-934EA
86.15
USD
102846-934
102846-936
3,3-Dimethyl-1-(trimethylsilyl)-1-butyne ≥97%
3,3-Dimethyl-1-(trimethylsilyl)-1-butyne
Hazard: Irritant
Formula:
C₉H₁₈Si |
MDL Number:
MFCD00134215 CAS Number: 14630-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...