3-[(3-Amino-3-oxopropyl)dithio]propanamide ≥97%
Supplier: Matrix Scientific
086093-1G
086093-10G
102846-902EA
276.92
USD
102846-902
102846-904
3-[(3-Amino-3-oxopropyl)dithio]propanamide ≥97%
3-[(3-Amino-3-oxopropyl)dithio]propanamide
Hazard: Irritant
Formula:
C₆H₁₂N₂O₂S₂ |
MDL Number:
MFCD00052129 CAS Number: 1002-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...