2-Methylpropanimidamide hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Isobutanamidine hydrochloride, Isobutyrimidamide hydrochloride

086092-1G 086092-10G
102846-898EA 127.69 USD
102846-898 102846-900
2-Methylpropanimidamide hydrochloride ≥97%
2-Methylpropanimidamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂
MW: 122.6 g/mol
MDL Number: MFCD00173787
CAS Number: 22007-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR