Dithiooxamide ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethanebis(thioamide), Rubeanic acid
086090-1G
086090-10G
086090-50G
102846-890EA
47.69
USD
102846-890
102846-892
102846-894
Dithiooxamide ≥97%
Dithiooxamide
Hazard: Irritant
Formula:
C₂H₄N₂S₂ |
MDL Number:
MFCD00004941 CAS Number: 79-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...