2,2,2-trifluoroethylhydrazine 65% in water , 97%

Supplier: Matrix Scientific

Synonyms: (2,2,2-Trifluoroethyl)hydrazine, 2,2,2-Trifluoroethylhydrazine

086087-1G 086087-10G
102846-884EA 156.92 USD
102846-884 102846-886
2,2,2-trifluoroethylhydrazine 65% in water , 97%
2,2,2-trifluoroethylhydrazine

Hazard: Irritant

Formula: C₂H₅F₃N₂
MDL Number: MFCD00007622
CAS Number: 5042-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR