Propagyl amine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride

086086-1G 086086-10G 086086-50G
102846-878EA 56.92 USD
102846-878 102846-880 102846-882
Propagyl amine hydrochloride ≥97%
Propagyl amine hydrochloride

Hazard: Irritant

Formula: C₃H₆ClN
MW: 91.54
MDL Number: MFCD00012907
CAS Number: 15430-52-1
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR