3-Buten-1-yl(trimethyl)silane ≥97%
Supplier: Matrix Scientific
086085-1G
086085-10G
102846-874EA
353.85
USD
102846-874
102846-876
3-Buten-1-yl(trimethyl)silane ≥97%
3-Buten-1-yl(trimethyl)silane
Purity: 97%
CAS: 763-13-3
MDL Number: MFCD00054880
Molecular Formula: C7H16Si
Molecular Weight: 128,29
Hazard: Irritant
Formula:
C₇H₁₆Si |
MDL Number:
MFCD00054880 CAS Number: 763-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...