Tris(dimethylamino)methane ≥90%
Supplier: Matrix Scientific
086084-1G
086084-10G
086084-50G
102846-868EA
175.38
USD
102846-868
102846-870
102846-872
Tris(dimethylamino)methane ≥90%
Tris(dimethylamino)methane
Hazard: Flammable/Corrosive/Refrigerate
Formula:
C₇H₁₉N₃ MW: 145.25 g/mol Boiling Pt: 42-43°C Density: 0.838 Flash Pt: 11°C |
MDL Number:
MFCD00008322 CAS Number: 5762-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...