3-Methylheptane ≥97%

Supplier: Matrix Scientific

086078-10G 086078-50G
102846-856EA 904.62 USD
102846-856 102846-858
3-Methylheptane ≥97%
3-Methylheptane

Hazard: Flammable

Formula: C₈H₁₈
Boiling Pt: 118…120 °C
Density: 0.71
Flash Pt: 7 °C
MDL Number: MFCD00027244
CAS Number: 589-81-1
UN: 1262
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR