Ethyl malonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl Malonyl Chloride

086075-25G 086075-100G
102846-850EA 127.69 USD
102846-850 102846-852
Ethyl malonyl chloride ≥95%
Ethyl malonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate/Lachrymator

Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 83 °C (22 mmHg)
Density: 1.189
Flash Pt: 67 °C
MDL Number: MFCD00000736
CAS Number: 36239-09-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR