N-Hydroxyurethane ≥97%

Supplier: Matrix Scientific

086073-1G 086073-10G 086073-50G
102846-844EA 72.31 USD
102846-844 102846-846 102846-848
N-Hydroxyurethane ≥97%
N-Hydroxyurethane

Hazard: Irritant

Formula: C₃H₇NO₃
MDL Number: MFCD00002108
CAS Number: 589-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR