2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate ≥97%
Supplier: Matrix Scientific
Synonyms:
DL-2,3-Dimercapto-1-propanesulphonic acid, sodium salt monohydrate, DMPS, Sodium 2,3-dimercaptopropane-1-sulphonate hydrate, 2,3-Dimercaptopropanesulphonic acid sodium salt, Sodium 2,3-Dimercaptopropanesulphonate monohydrate
086068-1G
086068-10G
102846-830EA
143.08
USD
102846-830
102846-832
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate ≥97%
2,3-Dimercapto-1-propanesulfonic acids odium salt monohydrate
Hazard: Irritant
Formula:
C₃H₉NaO₄S₃·H₂O MW: 228.30 g/mol (210.28 anhy) |
MDL Number:
MFCD00149543 CAS Number: 207233-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...