Cyanomethyl ethanethioate ≥97%

Supplier: Matrix Scientific

Synonyms: S-(Cyanomethyl) thioacetate

086064-1G 086064-10G
102846-816EA 338.46 USD
102846-816 102846-818
Cyanomethyl ethanethioate ≥97%
Cyanomethyl ethanethioate

Hazard: Irritant

Formula: C₄H₅NOS
MW: 115.15 g/mol
MDL Number: MFCD00173920
CAS Number: 59463-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR