4-Penten-1-ol ≥97%
Supplier: Matrix Scientific
Synonyms:
Pent-4-en-1-ol, 1-Penten-5-ol, 4-Penten-1-ol
086063-5G
086063-100G
102846-810EA
35.38
USD
102846-810
102846-814
4-Penten-1-ol ≥97%
4-Penten-1-ol
Hazard: Flammable
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 134…137 °C Density: 0.84 Flash Pt: 43 °C |
MDL Number:
MFCD00002975 CAS Number: 821-09-0 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...