1,1,1-Trifluoro-2,4-hexanedione ≥85%

Supplier: Matrix Scientific

086054-1G 086054-10G
102846-778EA 184.62 USD
102846-778 102846-780
1,1,1-Trifluoro-2,4-hexanedione ≥85%
1,1,1-Trifluoro-2,4-hexanedione

Hazard: Irritant

Formula: C₆H₇F₃O₂
MDL Number: MFCD00051306
CAS Number: 400-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR