N-(Cyclopropylmethyl)-N-propylamine ≥97%

Supplier: Matrix Scientific

086053-1G 086053-10G
102846-774EA 153.85 USD
102846-774 102846-776
N-(Cyclopropylmethyl)-N-propylamine ≥97%
N-(Cyclopropylmethyl)-N-propylamine

Hazard: Irritant

Formula: C₇H₁₅N
MW: 113.2 g/mol
MDL Number: MFCD00001308
CAS Number: 26389-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR