2-(Propylamino)ethanol ≥97%

Supplier: Matrix Scientific

086052-1G 086052-10G
102846-770EA 72.31 USD
102846-770 102846-772
2-(Propylamino)ethanol ≥97%
2-(Propylamino)ethanol

Hazard: Irritant

Formula: C₅H₁₃NO
MDL Number: MFCD00002844
CAS Number: 16369-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR