2-(Diallylamino)ethanol ≥97%

Supplier: Matrix Scientific

086050-1G 086050-10G
102846-762EA 1038.46 USD
102846-762 102846-764
2-(Diallylamino)ethanol ≥97%
2-(Diallylamino)ethanol

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD00039825
CAS Number: 17719-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR