Erbium trichloride hexahydrate ≥97%

Supplier: Matrix Scientific

086039-1G 086039-10G
102846-730EA 72.31 USD
102846-730 102846-732
Erbium trichloride hexahydrate ≥97%
Erbium trichloride hexahydrate

Purity: 97% CAS: 10025-75-9 MDL Number: MFCD00149693 Molecular Formula: Cl3ErH12O6 Molecular Weight: 381,71 Hazard: Irritant

Formula: Cl₃Er·6H₂O
MW: 381.71 g/mol
MDL Number: MFCD00149693
CAS Number: 10025-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR