4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol ≥97%
Supplier: Matrix Scientific
086037-1G
086037-10G
102846-722EA
127.69
USD
102846-722
102846-724
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol ≥97%
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol
Hazard: Irritant
Formula:
C₆H₇F₇O |
MDL Number:
MFCD00155992 CAS Number: 29819-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...