2-(Isopropylamino)ethanol ≥97%

Supplier: Matrix Scientific

Synonyms: N-Isopropylethanolamine

086028-10G 086028-50G
102846-690EA 76.92 USD
102846-690 102846-692
2-(Isopropylamino)ethanol ≥97%
2-(Isopropylamino)ethanol

Hazard: Irritant

Formula: C₅H₁₃NO
MW: 103.17 g/mol
Boiling Pt: 171 °C
Density: 0.9
Flash Pt: 76 °C
MDL Number: MFCD00041755
CAS Number: 109-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR