N,N'-Di-sec-butylcarbodiimide ≥98%
Supplier: Matrix Scientific
086021-1G
086021-10G
102846-668EA
255.38
USD
102846-668
102846-670
N,N'-Di-sec-butylcarbodiimide ≥98%
N,N'-Di-sec-butylcarbodiimide
Hazard: Irritant
Formula:
C₉H₁₈N₂ |
MDL Number:
MFCD01861858 CAS Number: 66006-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...