9-Fluorenone oxime ≥97%
Supplier: Matrix Scientific
Synonyms:
9H-Fluoren-9-one oxime
086005-1G
086005-10G
086005-50G
102846-620EA
72.31
USD
102846-620
102846-622
102846-624
9-Fluorenone oxime ≥97%
9-Fluorenone oxime
Hazard: Irritant
Formula:
C₁₃H₉NO |
MDL Number:
MFCD00016356 CAS Number: 2157-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...