6-Hydroxy-3-coumaranone ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one
085995-1G
085995-10G
102846-598EA
143.08
USD
102846-598
102846-600
6-Hydroxy-3-coumaranone ≥97%
6-Hydroxy-3-coumaranone
Hazard: Irritant
Formula:
C₈H₆O₃ MW: 150.14 g/mol |
MDL Number:
MFCD00068174 CAS Number: 6272-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...