Sodium-2-hydroxy-6-methoxy-1,2,3,4-tetrahydronaphthalene-2-sulfonate ≥97%
Supplier: Matrix Scientific
085993-1G
085993-10G
102846-592EA
340
USD
102846-592
102846-594
Sodium-2-hydroxy-6-methoxy-1,2,3,4-tetrahydronaphthalene-2-sulfonate ≥97%
Sodium-2-hydroxy-6-methoxy-1,2,3,4-tetrahydronaphthalene-2-sulfonate
Hazard: Irritant
Formula:
C₁₁H₁₃NaO₅S MW: 280.28 g/mol |
MDL Number:
MFCD05663953 CAS Number: 1017781-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...