Benzo[cd]indol-2(1H)-one ≥98%
Supplier: Matrix Scientific
Synonyms:
Naphthostyril, Naphtholactam, Benz[cd]indol-2(1H)-one
085969-10G
085969-50G
102846-520EA
516.92
USD
102846-520
102846-522
Benzo[cd]indol-2(1H)-one ≥98%
Benzo[cd]indol-2(1H)-one
Hazard: Irritant
Formula:
C₁₁H₇NO |
MDL Number:
MFCD00009748 CAS Number: 130-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...