1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone oxime ≥90%
Supplier: Matrix Scientific
085968-1G
085968-10G
102846-516EA
213.85
USD
102846-516
102846-518
1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone oxime ≥90%
1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone oxime
Hazard: Irritant
Formula:
C₁₂H₁₅NO Melting Pt: 103…104 °C |
MDL Number:
MFCD00099460 CAS Number: 7357-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...