N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine ≥97%

Supplier: Matrix Scientific

Synonyms: N-Fmoc-L-isoleucine, Fmoc-Ile-OH, FMOC-L-Isoleucine

085965-10G 085965-50G
102846-504EA 64.62 USD
102846-504 102846-506
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine ≥97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MDL Number: MFCD00037125
CAS Number: 71989-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR