N-(Chloromethyl)phthalimide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Chloromethyl)isoindoline-1,3-dione

085951-1G 085951-10G 085951-50G
102846-466EA 44.62 USD
102846-466 102846-468 102846-470
N-(Chloromethyl)phthalimide ≥97%
N-(Chloromethyl)phthalimide

Hazard: Irritant

Formula: C₉H₆ClNO₂
MDL Number: MFCD00005898
CAS Number: 17564-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR