N-{2-[2-(Acetylamino)phenyl]ethyl}-1-hydroxy-2-naphthamide ≥97%
Supplier: Matrix Scientific
085939-1G
085939-10G
102846-436EA
101.54
USD
102846-436
102846-438
N-{2-[2-(Acetylamino)phenyl]ethyl}-1-hydroxy-2-naphthamide ≥97%
N-{2-[2-(Acetylamino)phenyl]ethyl}-1-hydroxy-2-naphthamide
Hazard: Irritant
Formula:
C₂₁H₂₀N₂O₃ |
MDL Number:
MFCD00051399 CAS Number: 5254-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...