4-Methylphthalic anhydride ≥97%
Supplier: Matrix Scientific
085938-1G
085938-10G
102846-432EA
72.31
USD
102846-432
102846-434
4-Methylphthalic anhydride ≥97%
4-Methylphthalic anhydride
Hazard: Irritant
Formula:
C₉H₆O₃ MW: 162.14 g/mol |
MDL Number:
MFCD00041856 CAS Number: 19438-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...