2-(6-Bromo-2-methoxy-1-naphthyl)acetonitrile ≥97%

Supplier: Matrix Scientific

085931-1G 085931-10G
102846-416EA 353.85 USD
102846-416 102846-418
2-(6-Bromo-2-methoxy-1-naphthyl)acetonitrile ≥97%
2-(6-Bromo-2-methoxy-1-naphthyl)acetonitrile

Hazard: Irritant

Formula: C₁₃H₁₀BrNO
MDL Number: MFCD02180673
CAS Number: 92643-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR