6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-6,7-dimethoxyquinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-dione

085928-1G 085928-10G
102846-408EA 101.54 USD
102846-408 102846-410
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione ≥97%
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄
MW: 222.2 g/mol
MDL Number: MFCD00023889
CAS Number: 28888-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR