7-Methoxy-1-naphthylacetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
7-Methoxy-1-naphthylmethylcyanide
085917-1G
085917-5G
102846-378EA
461.54
USD
102846-378
102846-380
7-Methoxy-1-naphthylacetonitrile ≥97%
7-Methoxy-1-naphthylacetonitrile
Hazard: Irritant
Formula:
C₁₃H₁₁NO MW: 197.24 g/mol |
MDL Number:
MFCD08704309 CAS Number: 138113-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...