3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)benzo[d]thiazol-2(3H)-one

085912-1G 085912-10G
102846-364EA 241.54 USD
102846-364 102846-366
3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one ≥97%
3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one

Hazard: Irritant

Formula: C₈H₇NO₂S
MW: 181.21 g/mol
MDL Number: MFCD01074863
CAS Number: 72679-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR