(±)-1,2,3,4-Tetrahydro-1-naphthol ≥97%

Supplier: Matrix Scientific

Synonyms: ac-alpha-Tetralol, 1,2,3,4-Tetrahydronaphthalen-1-ol, 1,2,3,4-Tetrahydro-1-naphthol

085900-1G 085900-10G 085900-50G
102846-332EA 101.54 USD
102846-332 102846-334 102846-336
(±)-1,2,3,4-Tetrahydro-1-naphthol ≥97%
(±)-1,2,3,4-Tetrahydro-1-naphthol

Hazard: Irritant

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001739
CAS Number: 529-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR