3-Hydroxycoumarin ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-2-benzopyrone, 3-Hydroxy-2H-chromen-2-one
085897-1G
085897-10G
102846-322EA
184.62
USD
102846-322
102846-324
3-Hydroxycoumarin ≥97%
3-Hydroxycoumarin
Hazard: Irritant
Formula:
C₉H₆O₃ MW: 162.14 g/mol |
MDL Number:
MFCD00017490 CAS Number: 939-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...