Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥85%

Supplier: Matrix Scientific

Synonyms: Methyltetrahydrophthalic Anhydride

085896-1G 085896-10G
102846-318EA 72.31 USD
102846-318 102846-320
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥85%
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers)

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00014585
CAS Number: 11070-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR