1-Acetyl-4-fluoronaphthalene ≥97%

Supplier: Matrix Scientific

Synonyms: 4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone

085893-1G 085893-10G
102846-304EA 200 USD
102846-304 102846-306
1-Acetyl-4-fluoronaphthalene ≥97%
1-Acetyl-4-fluoronaphthalene

Hazard: Irritant

Formula: C₁₂H₉FO
MW: 188.20 g/mol
MDL Number: MFCD00134475
CAS Number: 316-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR