1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone ≥97%
Supplier: Matrix Scientific
085890-1G
085890-10G
102846-296EA
364.62
USD
102846-296
102846-298
1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone ≥97%
1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
Hazard: Irritant
Formula:
C₁₆H₂₂O |
MDL Number:
MFCD00052727 CAS Number: 17610-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...