6-Methoxy-2H-chromen-2-one ≥97%
Supplier: Matrix Scientific
085887-1G
085887-10G
102846-288EA
524.62
USD
102846-288
102846-290
6-Methoxy-2H-chromen-2-one ≥97%
6-Methoxy-2H-chromen-2-one
Hazard: Irritant
Formula:
C₁₀H₈O₃ |
MDL Number:
MFCD00016707 CAS Number: 17372-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...