7-Methyl-4-indanol ≥97%

Supplier: Matrix Scientific

Synonyms: 7-Methyl-2,3-dihydro-1H-inden-4-ol, 7-Methylindan-4-ol

085886-1G 085886-10G
102846-284EA 223.08 USD
102846-284 102846-286
7-Methyl-4-indanol ≥97%
7-Methyl-4-indanol

Hazard: Irritant

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00173675
CAS Number: 16400-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR